Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199351.1 | NP_001186280.1 | 9 - 10 | 999 | 56 | |
NM_001199352.1 | NP_001186281.1 | 9 - 10 | 863 | 56 | ||
NM_012040.3 | NP_036170.1 | 9 - 10 | 939 | 56 | ||
GenBank mRNA | AB023027.1 | - | 8 - 9 | 809 | 56 | |
AF181984.1 | - | 9 - 10 | 913 | 56 | ||
AK134665.1 | - | 9 - 10 | 870 | 56 | ||
AK158755.1 | - | 9 - 10 | 864 | 56 | ||
BC051996.1 | - | 9 - 10 | 846 | 56 | ||
BC055891.1 | - | 9 - 10 | 839 | 4208888 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |