Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013787.3 | NP_038815.1 | 1 - 2 | 274 | 90 | |
GenBank mRNA | AF083215.1 | - | 1 - 2 | 137 | 90 | |
AK032979.1 | - | 1 - 2 | 185 | 90 | ||
AK037002.1 | - | 1 - 2 | 173 | 90 | ||
AK044212.1 | - | 1 - 2 | 175 | 90 | ||
AK166034.1 | - | 1 - 2 | 175 | 90 | ||
AK167677.1 | - | 1 - 2 | 172 | 90 | ||
AK204645.1 | - | 1 - 2 | 143 | 90 | ||
BC003468.1 | - | 1 - 2 | 147 | 90 | ||
BC092236.1 | - | 1 - 2 | 148 | 30610417 | 90 | |
BY740905.1 | - | 1 - 2 | 275 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |