Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159284.1 | NP_001152756.1 | 9 - 10 | 1159 | 60 | |
NM_001284427.1 | NP_001271356.1 | 8 - 9 | 1108 | 60 | ||
NM_001284428.1 | NP_001271357.1 | 9 - 10 | 1216 | 60 | ||
NM_001284429.1 | NP_001271358.1 | 9 - 10 | 1161 | 60 | ||
NM_013870.3 | NP_038898.2 | 8 - 9 | 1108 | 60 | ||
GenBank mRNA | AF064236.1 | - | 8 - 9 | 1055 | 60 | |
AJ010305.1 | - | 8 - 9 | 1055 | 60 | ||
AK030932.1 | - | 8 - 9 | 1109 | 60 | ||
AK085412.1 | - | 9 - 10 | 1123 | 60 | ||
AK138898.1 | - | 9 - 10 | 1173 | 60 | ||
AK170516.1 | - | 8 - 9 | 1068 | 60 | ||
AK217205.1 | - | 1 - 2 | 86 | 60 | ||
BC010599.1 | - | 8 - 9 | 1049 | 3709078 | 60 | |
BC066192.1 | - | 9 - 10 | 1037 | 30433616 | 60 | |
BC069836.1 | - | 9 - 10 | 1037 | 30456568 | 60 | |
BC069839.1 | - | 9 - 10 | 1037 | 30431675 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |