Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013874.2 | NP_038902.1 | 4 - 5 | 498 | 59 | |
GenBank mRNA | AK142419.1 | - | 4 - 5 | 477 | 59 | |
BC052348.1 | - | 4 - 5 | 527 | 6587002 | 59 | |
U48238.1 | - | 4 - 5 | 497 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |