Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013875.5 | NP_038903.3 | 5 - 6 | 757 | 79 | |
GenBank mRNA | AF190639.1 | - | 5 - 6 | 538 | 79 | |
AJ251859.1 | - | 5 - 6 | 809 | 79 | ||
AK035385.1 | - | 5 - 6 | 769 | 79 | ||
AK049910.1 | - | 5 - 6 | 813 | 79 | ||
BC130267.1 | - | 5 - 6 | 638 | 40143060 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |