Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scmh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159630.1 | NP_001153102.1 | 11 - 12 | 1534 | 60 | |
NM_013883.2 | NP_038911.1 | 11 - 12 | 1534 | 60 | ||
GenBank mRNA | AB030906.1 | - | 11 - 12 | 1433 | 60 | |
AB221680.1 | - | 7 - 8 | 1075 | 60 | ||
AB221681.1 | - | 7 - 8 | 1075 | 60 | ||
BC025478.1 | - | 2 - 3 | 1776 | 60 | ||
BC034667.1 | - | 9 - 10 | 1244 | 5356322 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |