Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scmh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159630.1 | NP_001153102.1 | 14 - 15 | 2122 | 87 | |
NM_013883.2 | NP_038911.1 | 14 - 15 | 2122 | 87 | ||
GenBank mRNA | AB030906.1 | - | 14 - 15 | 2021 | 87 | |
AB221680.1 | - | 10 - 11 | 1663 | 87 | ||
AB221681.1 | - | 10 - 11 | 1663 | 87 | ||
AK009416.1 | - | 2 - 3 | 280 | 87 | ||
BC034667.1 | - | 12 - 13 | 1832 | 5356322 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |