Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016777.3 | NP_058057.3 | 6 - 7 | 511 | 114 | |
GenBank mRNA | AF034610.2 | - | 6 - 7 | 501 | 114 | |
AK132690.1 | - | 6 - 7 | 490 | 114 | ||
AK161367.1 | - | 6 - 7 | 490 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |