Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016910.3 | NP_058606.3 | 4 - 5 | 1212 | 66 | |
GenBank mRNA | AF200464.1 | - | 4 - 5 | 993 | 66 | |
AK157818.1 | - | 4 - 5 | 1200 | 66 | ||
AK171714.1 | - | 4 - 5 | 1192 | 66 | ||
AK181083.1 | - | 1 - 2 | 105 | 66 | ||
BC023492.1 | - | 4 - 5 | 1088 | 5026376 | 66 | |
BC051966.1 | - | 4 - 5 | 1212 | 5689560 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |