Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016964.2 | NP_058660.2 | 4 - 5 | 534 | 95 | |
GenBank mRNA | AJ005678.1 | - | 4 - 5 | 492 | 95 | |
AK145898.1 | - | 4 - 5 | 520 | 95 | ||
AK158240.1 | - | 4 - 5 | 430 | 95 | ||
BC060184.1 | - | 4 - 5 | 534 | 95 | ||
BC139010.1 | - | 4 - 5 | 464 | 8862032 | 95 | |
BC139011.1 | - | 4 - 5 | 464 | 8862033 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |