Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016964.2 | NP_058660.2 | 14 - 15 | 1670 | 95 | |
GenBank mRNA | AJ005678.1 | - | 14 - 15 | 1628 | 95 | |
AK145898.1 | - | 13 - 14 | 2157 | 95 | ||
BC060184.1 | - | 13 - 14 | 2111 | 95 | ||
BC139010.1 | - | 14 - 15 | 1600 | 8862032 | 95 | |
BC139011.1 | - | 14 - 15 | 1600 | 8862033 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |