Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017375.3 | NP_059071.1 | 2 - 3 | 242 | 116 | |
GenBank mRNA | AK002899.1 | - | 2 - 3 | 221 | 116 | |
AK005525.1 | - | 2 - 3 | 211 | 116 | ||
AK008123.1 | - | 2 - 3 | 190 | 116 | ||
AK008467.1 | - | 2 - 3 | 190 | 116 | ||
AK010074.1 | - | 2 - 3 | 238 | 116 | ||
AK145949.1 | - | 2 - 3 | 193 | 116 | ||
AK149175.1 | - | 220 | 116 | |||
AK150145.1 | - | 2 - 3 | 212 | 116 | ||
AK155028.1 | - | 2 - 3 | 212 | 116 | ||
BC060986.1 | - | 2 - 3 | 121 | 30300544 | 116 | |
BY065904.1 | - | 2 - 3 | 242 | 116 | ||
U58888.1 | - | 2 - 3 | 190 | 116 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BC020352.1 | - | Chr. - - [-] on Build | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |