Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017395.2 | NP_059091.2 | 6 - 7 | 573 | 61 | |
GenBank mRNA | AB221679.1 | - | 6 - 7 | 554 | 61 | |
AF209854.1 | - | 6 - 7 | 554 | 61 | ||
AK088686.1 | - | 6 - 7 | 739 | 61 | ||
BC051965.1 | - | 7 - 8 | 610 | 5702640 | 61 | |
BC057336.1 | - | 7 - 8 | 599 | 61 | ||
BC075717.1 | - | 8 - 9 | 682 | 61 | ||
BC118948.2 | - | 6 - 7 | 554 | 40092396 | 61 | |
BC118949.2 | - | 6 - 7 | 554 | 40092397 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |