Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400001538.mm.1 |
NM_001205241.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001538.mm.1 |
NM_001205241.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001538.mm.1 |
NM_017479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001538.mm.1 |
NM_017479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001538.mm.1 |
TC1400000195.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001541.mm.1 |
NM_001205241.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001541.mm.1 |
NM_001205241.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001541.mm.1 |
NM_017479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001541.mm.1 |
NM_017479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001541.mm.1 |
TC1400000195.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001536.mm.1 |
NM_001205241.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001536.mm.1 |
NM_001205241.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001536.mm.1 |
NM_017479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001536.mm.1 |
NM_017479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001536.mm.1 |
TC1400000195.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001539.mm.1 |
NM_001205241.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001539.mm.1 |
NM_001205241.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001539.mm.1 |
NM_017479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001539.mm.1 |
NM_017479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001539.mm.1 |
TC1400000195.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001537.mm.1 |
NM_001205241.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001537.mm.1 |
NM_001205241.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001537.mm.1 |
NM_017479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001537.mm.1 |
NM_017479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001537.mm.1 |
TC1400000195.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001542.mm.1 |
NM_001205241.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001542.mm.1 |
NM_001205241.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001542.mm.1 |
NM_017479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001542.mm.1 |
NM_017479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001542.mm.1 |
TC1400000195.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001540.mm.1 |
NM_001205241.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001540.mm.1 |
NM_001205241.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001540.mm.1 |
NM_017479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001540.mm.1 |
NM_017479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001540.mm.1 |
TC1400000195.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001535.mm.1 |
NM_001205241.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001535.mm.1 |
NM_001205241.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001535.mm.1 |
NM_017479.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001535.mm.1 |
NM_017479.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001535.mm.1 |
TC1400000195.mm.1 |
MTA TC |