Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018889.3 | NP_061377.2 | 4 - 5 | 595 | 82 | |
GenBank mRNA | AK153819.1 | - | 4 - 5 | 512 | 82 | |
BC052658.1 | - | 4 - 5 | 555 | 30048340 | 82 | |
D84436.1 | - | 4 - 5 | 595 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |