Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019645.2 | NP_062619.1 | 7 - 8 | 1369 | 90 | |
GenBank mRNA | AK036856.1 | - | 7 - 8 | 1554 | 90 | |
AK079429.1 | - | 7 - 8 | 1554 | 90 | ||
BC139041.1 | - | 7 - 8 | 1464 | 8862063 | 90 | |
BC139042.1 | - | 7 - 8 | 1465 | 8862064 | 90 | |
Y07941.1 | - | 7 - 8 | 1369 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |