Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019656.3 | NP_062630.2 | 4 - 5 | 584 | 152 | |
GenBank mRNA | AF053454.1 | - | 4 - 5 | 531 | 152 | |
AK146673.1 | - | 4 - 5 | 489 | 152 | ||
AK149204.1 | - | 4 - 5 | 494 | 152 | ||
AK160313.1 | - | 4 - 5 | 478 | 152 | ||
AK167384.1 | - | 4 - 5 | 480 | 152 | ||
BC003733.1 | - | 4 - 5 | 518 | 3590862 | 152 | |
BC110314.1 | - | 4 - 5 | 493 | 5344878 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |