Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019810.4 | NP_062784.3 | 6 - 7 | 819 | 74 | |
GenBank mRNA | AF163846.1 | - | 6 - 7 | 581 | 74 | |
AF208031.1 | - | 6 - 7 | 819 | 74 | ||
AK033512.1 | - | 6 - 7 | 805 | 74 | ||
AK085602.1 | - | 6 - 7 | 610 | 74 | ||
BC003845.1 | - | 6 - 7 | 799 | 3497611 | 74 | |
U27269.1 | - | 4 - 5 | 389 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |