Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019911.2 | NP_064295.2 | 1 - 2 | 63 | 81 | |
GenBank mRNA | AB476989.1 | - | 1 - 2 | 537 | 81 | |
AK132271.1 | - | 1 - 2 | 86 | 81 | ||
BC018390.1 | - | 1 - 2 | 63 | 4159877 | 81 | |
U24493.1 | - | 1 - 2 | 50 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |