Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000012920.mm.1 |
NM_001164195.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012920.mm.1 |
NM_001164195.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012920.mm.1 |
NM_019961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012920.mm.1 |
NM_019961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012920.mm.1 |
TC1000001772.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012926.mm.1 |
NM_001164195.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012926.mm.1 |
NM_001164195.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012926.mm.1 |
NM_019961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012926.mm.1 |
NM_019961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012926.mm.1 |
TC1000001772.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012929.mm.1 |
NM_001164195.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012929.mm.1 |
NM_001164195.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012929.mm.1 |
NM_019961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012929.mm.1 |
NM_019961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012929.mm.1 |
TC1000001772.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012927.mm.1 |
NM_001164195.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012927.mm.1 |
NM_001164195.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012927.mm.1 |
NM_019961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012927.mm.1 |
NM_019961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012927.mm.1 |
TC1000001772.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012928.mm.1 |
NM_001164195.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012928.mm.1 |
NM_001164195.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012928.mm.1 |
NM_019961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012928.mm.1 |
NM_019961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012928.mm.1 |
TC1000001772.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012925.mm.1 |
NM_001164195.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012925.mm.1 |
NM_001164195.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012925.mm.1 |
NM_019961.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012925.mm.1 |
NM_019961.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012925.mm.1 |
TC1000001772.mm.1 |
MTA TC |