Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164621.1 | NP_001158093.1 | 5 - 6 | 959 | 73 | |
NM_020012.2 | NP_064396.2 | 4 - 5 | 646 | 73 | ||
GenBank mRNA | AF249667.1 | - | 4 - 5 | 592 | 73 | |
AK011316.2 | - | 5 - 6 | 647 | 73 | ||
AK195837.1 | - | 1 - 2 | 150 | 73 | ||
BC049079.1 | - | 5 - 6 | 959 | 73 | ||
BC054841.1 | - | 4 - 5 | 576 | 6489903 | 73 | |
BC094250.1 | - | 4 - 5 | 597 | 5720753 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |