Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205085.1 | NP_001192014.1 | 3 - 4 | 999 | 77 | |
NM_020496.3 | NP_065242.1 | 3 - 4 | 999 | 77 | ||
NM_194263.2 | NP_919239.1 | 3 - 4 | 999 | 77 | ||
GenBank mRNA | AF260557.1 | - | 3 - 4 | 1004 | 77 | |
AF306667.1 | - | 3 - 4 | 937 | 77 | ||
AJ277486.1 | - | 3 - 4 | 541 | 77 | ||
AK052155.1 | - | 3 - 4 | 981 | 77 | ||
AK052389.1 | - | 3 - 4 | 982 | 77 | ||
AK141251.1 | - | 3 - 4 | 769 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |