Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021301.3 | NP_067276.2 | 14 - 15 | 1406 | 107 | |
GenBank mRNA | AF111811.1 | - | 14 - 15 | 1406 | 107 | |
AK128939.1 | - | 14 - 15 | 1239 | 107 | ||
BC138627.1 | - | 14 - 15 | 1363 | 8861647 | 107 | |
BC138628.1 | - | 14 - 15 | 1363 | 8861648 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |