Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025571.2 | NP_001020742.1 | 1 - 2 | 355 | 81 | |
NM_021309.3 | NP_067284.1 | 1 - 2 | 355 | 81 | ||
GenBank mRNA | AF203343.1 | - | 1 - 2 | 362 | 81 | |
AK037489.1 | - | 1 - 2 | 77 | 81 | ||
AK172182.1 | - | 1 - 2 | 107 | 81 | ||
BB631086.1 | - | 1 - 2 | 99 | 81 | ||
BC034847.1 | - | 1 - 2 | 87 | 3327113 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |