Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piwil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021311.3 | NP_067286.1 | 11 - 12 | 1361 | 84 | |
GenBank mRNA | AB032604.1 | - | 11 - 12 | 1326 | 84 | |
AF438405.1 | - | 11 - 12 | 1361 | 84 | ||
BC066846.1 | - | 1412 | 84 | |||
BC129857.1 | - | 10 - 11 | 1211 | 40109791 | 84 | |
BC129858.1 | - | 10 - 11 | 1211 | 40109793 | 84 | |
EF196092.1 | - | 9 - 10 | 1171 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |