Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngly1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021504.3 | NP_067479.2 | 1 - 2 | 183 | 71 | |
GenBank mRNA | AF250927.1 | - | 1 - 2 | 170 | 71 | |
AK028248.1 | - | 1 - 2 | 184 | 71 | ||
AK169205.1 | - | 1 - 2 | 171 | 71 | ||
BC028961.1 | - | 1 - 2 | 137 | 5366023 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |