Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253860.1 | NP_001240789.1 | 4 - 5 | 668 | 70 | |
GenBank mRNA | AJ271477.1 | - | 4 - 5 | 664 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Ion Channel Mus musculus Probe spans exons |