Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022411.3 | NP_071856.1 | 8 - 9 | 1185 | 63 | |
GenBank mRNA | AF201903.1 | - | 8 - 9 | 1185 | 63 | |
AK140972.1 | - | 8 - 9 | 1236 | 63 | ||
BC013493.1 | - | 8 - 9 | 1210 | 4218337 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |