Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122765.1 | NP_001116237.1 | 5 - 6 | 423 | 92 | |
NM_001122766.1 | NP_001116238.1 | 3 - 4 | 260 | 92 | ||
NM_022432.4 | NP_071877.3 | 6 - 7 | 470 | 92 | ||
GenBank mRNA | AB221570.1 | - | 6 - 7 | 375 | 92 | |
AF299337.1 | - | 6 - 7 | 455 | 92 | ||
AK014042.1 | - | 5 - 6 | 394 | 92 | ||
AK144431.1 | - | 5 - 6 | 366 | 92 | ||
AK146412.1 | - | 5 - 6 | 403 | 92 | ||
BB868649.1 | - | 3 - 4 | 225 | 92 | ||
BC021439.1 | - | 6 - 7 | 474 | 5039278 | 92 | |
KF032392.1 | - | 6 - 7 | 412 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |