Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023141.2 | NP_075630.2 | 1 - 2 | 251 | 70 | |
GenBank mRNA | AJ299404.1 | - | 1 - 2 | 219 | 70 | |
AK007814.1 | - | 1 - 2 | 240 | 70 | ||
AK009693.1 | - | 1 - 2 | 269 | 70 | ||
AK151048.1 | - | 1 - 2 | 237 | 70 | ||
AK151390.1 | - | 1 - 2 | 218 | 70 | ||
AK152243.1 | - | 1 - 2 | 216 | 70 | ||
AK152358.1 | - | 1 - 2 | 252 | 70 | ||
AK152484.1 | - | 1 - 2 | 252 | 70 | ||
AK153391.1 | - | 1 - 2 | 237 | 70 | ||
AK170296.1 | - | 1 - 2 | 246 | 70 | ||
AK172355.1 | - | 1 - 2 | 226 | 70 | ||
BC025525.1 | - | 1 - 2 | 236 | 70 | ||
BC052851.1 | - | 1 - 2 | 249 | 70 | ||
BC071216.1 | - | 1 - 2 | 232 | 30439199 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |