Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025302.3 | NP_079578.1 | 4 - 5 | 583 | 83 | |
GenBank mRNA | AK009521.1 | - | 4 - 5 | 577 | 83 | |
BC027762.1 | - | 4 - 5 | 584 | 5353459 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |