Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025835.2 | NP_080111.1 | 1 - 2 | 209 | 102 | |
GenBank mRNA | AF327060.1 | - | 1 - 2 | 197 | 102 | |
AK004976.1 | - | 1 - 2 | 210 | 102 | ||
AK050366.1 | - | 1 - 2 | 215 | 102 | ||
AK134437.1 | - | 1 - 2 | 220 | 102 | ||
BC026531.1 | - | 1 - 2 | 185 | 102 | ||
BC037082.1 | - | 1 - 2 | 211 | 5098790 | 102 | |
BC055946.1 | - | 1 - 2 | 216 | 4166700 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |