Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026141.3 | NP_080417.2 | 5 - 6 | 513 | 125 | |
GenBank mRNA | AK011443.1 | - | 5 - 6 | 485 | 125 | |
AK012371.1 | - | 5 - 6 | 490 | 125 | ||
AK013377.1 | - | 5 - 6 | 486 | 125 | ||
AK014406.2 | - | 486 | 125 | |||
AK014453.1 | - | 4 - 5 | 1038 | 125 | ||
AK017370.1 | - | 5 - 6 | 513 | 125 | ||
AK167168.1 | - | 5 - 6 | 478 | 125 | ||
AK167258.1 | - | 5 - 6 | 485 | 125 | ||
BC079912.1 | - | 5 - 6 | 475 | 30661438 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |