Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253712.1 | NP_001240641.1 | 3 - 4 | 516 | 69 | |
NM_001253714.1 | NP_001240643.1 | 2 - 3 | 433 | 69 | ||
NM_001253716.1 | NP_001240645.1 | 3 - 4 | 244 | 69 | ||
NM_026322.4 | NP_080598.2 | 8 - 9 | 915 | 69 | ||
GenBank mRNA | AK002356.1 | - | 3 - 4 | 391 | 69 | |
AK009822.1 | - | 3 - 4 | 393 | 69 | ||
AK018338.1 | - | 3 - 4 | 238 | 69 | ||
AK049714.1 | - | 3 - 4 | 517 | 69 | ||
BC089311.1 | - | 8 - 9 | 915 | 6412625 | 69 | |
CB953198.1 | - | 2 - 3 | 304 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |