Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289535.1 | NP_001276464.1 | 1 - 2 | 265 | 68 | |
NM_001289536.1 | NP_001276465.1 | 1 - 2 | 265 | 68 | ||
NM_026959.3 | NP_081235.2 | 1 - 2 | 265 | 68 | ||
GenBank mRNA | AK007700.1 | - | 1 - 2 | 263 | 68 | |
AK016910.1 | - | 1 - 2 | 207 | 68 | ||
AK169273.1 | - | 1 - 2 | 209 | 68 | ||
AK170165.1 | - | 1 - 2 | 261 | 68 | ||
BC021362.1 | - | 1 - 2 | 255 | 5035350 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |