Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcolce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029620.2 | NP_083896.1 | 5 - 6 | 878 | 135 | |
GenBank mRNA | AF352788.1 | - | 5 - 6 | 774 | 135 | |
AK010249.1 | - | 5 - 6 | 879 | 135 | ||
AK132395.1 | - | 768 | 135 | |||
BC051174.1 | - | 5 - 6 | 751 | 6310294 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |