Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271725.1 | NP_001258654.1 | 4 - 5 | 725 | 93 | |
NM_001271726.1 | NP_001258655.1 | 4 - 5 | 761 | 93 | ||
NM_001271727.1 | NP_001258656.1 | 4 - 5 | 1030 | 93 | ||
NM_001271728.1 | NP_001258657.1 | 4 - 5 | 828 | 93 | ||
NM_030706.3 | NP_109631.2 | 4 - 5 | 824 | 93 | ||
GenBank mRNA | AB043550.1 | - | 4 - 5 | 808 | 93 | |
AF220017.1 | - | 4 - 5 | 673 | 93 | ||
AK039741.1 | - | 4 - 5 | 822 | 93 | ||
AK045410.1 | - | 4 - 5 | 821 | 93 | ||
AK079415.1 | - | 4 - 5 | 975 | 93 | ||
AK141419.1 | - | 4 - 5 | 671 | 93 | ||
AK147275.1 | - | 4 - 5 | 938 | 93 | ||
AK147395.1 | - | 4 - 5 | 819 | 93 | ||
AK164350.1 | - | 3 - 4 | 527 | 93 | ||
AK220343.1 | - | 4 - 5 | 1029 | 93 | ||
BC058668.1 | - | 4 - 5 | 792 | 93 | ||
BC058961.1 | - | 4 - 5 | 828 | 6826143 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |