Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031201.1 | NP_112478.1 | 4 - 5 | 481 | 73 | |
GenBank mRNA | AK029632.1 | - | 4 - 5 | 505 | 73 | |
BC089003.1 | - | 3 - 4 | 417 | 6818427 | 73 | |
M30127.2 | - | 4 - 5 | 481 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |