Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172098.1 | NP_001165569.1 | 11 - 12 | 1679 | 92 | |
NM_052973.2 | NP_443205.1 | 13 - 14 | 1931 | 92 | ||
GenBank mRNA | AF307777.1 | - | 13 - 14 | 1915 | 92 | |
AK140447.1 | - | 11 - 12 | 1667 | 92 | ||
AK165908.1 | - | 11 - 12 | 1363 | 92 | ||
BC138055.1 | - | 11 - 12 | 1630 | 8861074 | 92 | |
BC145127.1 | - | 11 - 12 | 1630 | 9053675 | 92 | |
EF685152.1 | - | 14 - 15 | 1832 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |