Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053204.2 | NP_444434.2 | 9 - 10 | 2303 | 94 | |
NR_104484.1 | 9 - 10 | 2303 | 94 | |||
GenBank mRNA | AF340028.1 | - | 9 - 10 | 2287 | 94 | |
AF340029.1 | - | 9 - 10 | 2287 | 94 | ||
AY316692.1 | - | 7 - 8 | 1735 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |