Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rims2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256382.1 | NP_001243311.1 | 25 - 26 | 4437 | 75 | |
NM_001256383.1 | NP_001243312.1 | 24 - 25 | 4371 | 75 | ||
NM_001256384.1 | NP_001243313.1 | 4 - 5 | 773 | 75 | ||
NM_053271.2 | NP_444501.1 | 25 - 26 | 4311 | 75 | ||
GenBank mRNA | AB021131.1 | - | 25 - 26 | 4265 | 75 | |
AK032619.1 | - | 4 - 5 | 719 | 75 | ||
AK129206.1 | - | 22 - 23 | 3535 | 75 | ||
BC118979.2 | - | 24 - 25 | 4319 | 40098740 | 75 | |
HM015529.1 | - | 25 - 26 | 4360 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |