Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Recql4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058214.3 | NP_478121.2 | 18 - 19 | 3167 | 82 | |
GenBank mRNA | AB039882.1 | - | 17 - 18 | 3193 | 82 | |
AB175741.1 | - | 18 - 19 | 3084 | 82 | ||
AK090260.1 | - | 18 - 19 | 2848 | 82 | ||
BC064680.1 | - | 18 - 19 | 3141 | 82 | ||
BC131656.1 | - | 18 - 19 | 3080 | 82 | ||
BC131657.1 | - | 16 - 17 | 3417 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |