Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024856.2 | NP_001020027.1 | 7 - 8 | 1035 | 82 | |
NM_001024857.2 | NP_001020028.1 | 6 - 7 | 961 | 82 | ||
NM_080788.3 | NP_542966.2 | 7 - 8 | 789 | 82 | ||
GenBank mRNA | AB046593.2 | - | 6 - 7 | 713 | 82 | |
AK046978.1 | - | 7 - 8 | 1008 | 82 | ||
AK053820.1 | - | 7 - 8 | 1024 | 82 | ||
AK077011.1 | - | 7 - 8 | 810 | 82 | ||
AK137024.1 | - | 6 - 7 | 950 | 82 | ||
AK148269.1 | - | 6 - 7 | 965 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mus musculus Probe spans exons |