Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290651.1 | NP_001277580.1 | 11 - 12 | 1581 | 63 | |
NM_001290652.1 | NP_001277581.1 | 11 - 12 | 1581 | 63 | ||
GenBank mRNA | AF285577.1 | - | 13 - 14 | 1407 | 63 | |
AK016533.1 | - | 14 - 15 | 1453 | 63 | ||
AK031490.1 | - | 11 - 12 | 1560 | 63 | ||
AK040521.1 | - | 11 - 12 | 1583 | 63 | ||
BC056382.1 | - | 11 - 12 | 1268 | 6853936 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |