Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133241.2 | NP_573504.1 | 7 - 8 | 650 | 77 | |
GenBank mRNA | AF449425.1 | - | 7 - 8 | 1104 | 77 | |
AK030396.1 | - | 7 - 8 | 649 | 77 | ||
AK043757.1 | - | 7 - 8 | 650 | 77 | ||
AK122190.1 | - | 7 - 8 | 1092 | 77 | ||
BC024719.1 | - | 7 - 8 | 650 | 5363238 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |