Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286007.1 | NP_001272936.1 | 7 - 8 | 1389 | 59 | |
NM_001286009.1 | NP_001272938.1 | 7 - 8 | 1389 | 59 | ||
NM_001286011.1 | NP_001272940.1 | 6 - 7 | 1156 | 59 | ||
NM_133755.2 | NP_598516.2 | 7 - 8 | 1316 | 59 | ||
GenBank mRNA | AK033157.1 | - | 7 - 8 | 1106 | 59 | |
AK045084.1 | - | 7 - 8 | 1105 | 59 | ||
AK146771.1 | - | 7 - 8 | 1311 | 59 | ||
AK153511.1 | - | 7 - 8 | 1237 | 59 | ||
AK158060.1 | - | 7 - 8 | 1105 | 59 | ||
AK167924.1 | - | 7 - 8 | 1211 | 59 | ||
AK194085.1 | - | 2 - 3 | 225 | 59 | ||
BC012519.1 | - | 7 - 8 | 1244 | 3601589 | 59 | |
BC025564.1 | - | 6 - 7 | 1078 | 59 | ||
BC025582.1 | - | 7 - 8 | 1205 | 5324981 | 59 | |
BC027034.1 | - | 7 - 8 | 1300 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |