Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134064.2 | NP_598825.2 | 4 - 5 | 762 | 89 | |
NR_027396.1 | 4 - 5 | 679 | 89 | |||
GenBank mRNA | AK160090.1 | - | 4 - 5 | 613 | 89 | |
AK160115.1 | - | 4 - 5 | 613 | 89 | ||
BC017630.1 | - | 4 - 5 | 555 | 4507289 | 89 | |
BC035548.1 | - | 4 - 5 | 577 | 4221284 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |