Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144868.3 | NP_659117.2 | 10 - 11 | 2990 | 79 | |
GenBank mRNA | AF237953.1 | - | 8 - 9 | 1007 | 79 | |
BC038056.1 | - | 5 - 6 | 1098 | 79 | ||
BC054751.1 | - | 10 - 11 | 2269 | 79 | ||
BC055289.1 | - | 11 - 12 | 2793 | 79 | ||
BC096538.1 | - | 10 - 11 | 2990 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |