Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144868.3 | NP_659117.2 | 14 - 15 | 3471 | 77 | |
GenBank mRNA | AF237953.1 | - | 12 - 13 | 1488 | 77 | |
BC025838.1 | - | 8 - 9 | 974 | 77 | ||
BC038056.1 | - | 9 - 10 | 1579 | 77 | ||
BC054751.1 | - | 14 - 15 | 2750 | 77 | ||
BC055289.1 | - | 15 - 16 | 3274 | 77 | ||
BC096538.1 | - | 14 - 15 | 3471 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |