Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144868.3 | NP_659117.2 | 17 - 18 | 3785 | 94 | |
GenBank mRNA | AF237953.1 | - | 15 - 16 | 1802 | 94 | |
BC025838.1 | - | 11 - 12 | 1288 | 94 | ||
BC038056.1 | - | 12 - 13 | 1893 | 94 | ||
BC054751.1 | - | 17 - 18 | 3064 | 94 | ||
BC055289.1 | - | 18 - 19 | 3588 | 94 | ||
BC096538.1 | - | 17 - 18 | 3785 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |